Skip to Content
MilliporeSigma
All Photos(29)

Documents

ZRB1648

Sigma-Aldrich

Anti-ATG9A Antibody, clone 5B11, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

APG9-like 1, Autophagy-related protein 9A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5B11, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 94.45 kDa
observed mol wt ~110 kDa

species reactivity

human

species reactivity (predicted by homology)

canine, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10-1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5B11 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Autophagy-related protein 9A. It targets an epitope with in 16 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human Autophagy-related protein 9A.

Application

Anti-ATG9A, clone 5B11 ZooMAb, Cat. No. ZRB1648, is a recombinant Rabbit Monoclonal Antibody that specifically targets Autophagy-related protein 9A and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ATG9A in HepG2 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:10 dilution from a representative lot detected ATG9A in human spleen and human kidney tissue sections.

Immunocytochemistry Analysis: A 1:1000 dilution from a representative lot detected ATG9A in HepG2 cells.

Target description

Autophagy-related protein 9A (UniProt: Q7Z3C6; also known as APG9-like 1, ATG9) is encoded by the ATG9A (also known as APG9L1) gene (Gene ID: 79065) in human. ATG9A is a multi-pass membrane protein that is involved in autophagy and cytoplasm to vacuole transport vesicle formation. It plays a key role in the organization of the preautophagosomal structure/phagophore assembly site. Among core autophagy-related proteins, ATG9A is the only multi-spanning transmembrane protein. Both its N- and C-terminal domains are exposed to the cytoplasm. Its N-terminal region (aa 1-36) is important in its role in autophagosome formation and this region is required for both autophagy and its Golgi localization. This region also contains sorting motifs known as the di-acidic motif (aa 5-7), a tyrosine-based motif (aa 8-11), and a di-leucine-based motif (aa 22-27). In mammalian cells, ATG9A is mainly localized to the trans-Golgi network and the endosomal system. It is shown to cycle between them though vesicle trafficking. ATG9A travels through the trans-Golgi network and the endosomal system under nutrient-rich conditions, however, upon induction of autophagy it transiently localizes to the autophagosome. Under amino acid starvation or upon rapamycin treatment, it is shown to redistribute from a juxtanuclear clustered pool to a dispersed peripheral cytosolic pool. Mutant forms of ATG9A that lack both the tyrosine-based and di-leucine-based sorting motifs exhibit reduced Golgi localization and abnormal accumulation on the recycling endosomes under conditions of nutrient deprivation. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Imai, K., et al. (2016). J. Cell Sci. 129(20); 3781-3791).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Belinda S Hall et al.
Autophagy, 18(4), 841-859 (2021-08-24)
The Mycobacterium ulcerans exotoxin, mycolactone, is responsible for the immunosuppression and tissue necrosis that characterizes Buruli ulcer. Mycolactone inhibits SEC61-dependent co-translational translocation of proteins into the endoplasmic reticulum and the resultant cytosolic translation triggers degradation of mislocalized proteins by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service