Skip to Content
MilliporeSigma
All Photos(16)

Key Documents

ZRB1547

Sigma-Aldrich

Anti-phospho-HDAC4 (S246) Antibody, clone 1I14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Histone deacetylase 4;EC:3.5.1.98;HD4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I14, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 119.04 and 106.37 kDa
observed mol wt ~120 and 110 kDa

purified by

using Protein A

species reactivity

human, rat

species reactivity (predicted by homology)

bovine, monkey, canine, feline, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer246)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I14 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Histone deacetylase 4 (HDAC4) phosphorylated on serine 246. It detects both isoforms of HDAC4.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 246 from the N-terminal half of human HDAC4.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells treated with EGF.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected HDAC4 phosphorylated on serine 246 in lysate from overnight serum starved HEK293 cells treated with recombinant human EGF (50 ng/mL for 10 min).

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-HDAC4 (S246) in PC12 treated with EGF.

Affinity Binding Assay: A representative lot of this antibody bound phospho-HDAC4 (Ser246) with a KD of 7.8 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-HDAC4 (S246) in human breast cancer tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-HDAC4 (S246) in HEK293 cells treated with EGF.

Peptide Inhibition Assay: Target band detection in lysate from HEK293 cells treated with EGF was prevented by preblocking of a representative lot with phospho-HDAC4 peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Histone deacetylase 4 (UniProt: P56524; also known as EC:3.5.1.98, HD4) is encoded by the HDAC4 (also known as KIAA0288) gene (Gene ID: 9759) in human. HDAC4 is a ubiquitous enzyme that is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression, and developmental events. It is present as a homodimeric protein that shuttles between the nucleus and the cytoplasm. Its nuclear export sequence is localized in amino acids 1051-1084. Its export to cytoplasm depends on the interaction with a 14-3-3 chaperone protein. 14-3-3 family of proteins are known to bind specifically to conserved phosphoserine-containing motifs. Phosphorylation of specific amino acids creates binding sites for the 14-3-3, which escorts phospho- HDAC4 from the nucleus to the cytoplasm. Phosphorylation of HDAC4 at serine 246, 467, and 632 is achieved by CaMK4. Its nuclear localization probably depends on sumoylation on lysine 559, which is promoted by E3 SUMO-protein ligase RanBP2. Two isoforms of HDAC4 have been described that are produced by alternative splicing. Isoform 2 lacks the first 117 amino acids. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wang, Z., et al. (2014). Epigenomics 6(1); 139-150; Wang, AH., et al. (2000). Mol. Cell Biol. 20(18); 6904-6912).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service