Skip to Content
MilliporeSigma
All Photos(13)

Key Documents

ZRB1450

Sigma-Aldrich

Anti-PIAS Antibody, clone 1O17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

DEAD/H box-binding protein 1, E3 SUMO-protein ligase PIAS1, E3 SUMO-protein transferase PIAS1, GBP, Gu-binding protein, Protein inhibitor of activated STAT protein 1, RNA helicase II-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O17, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 71.84 kDa
observed mol wt 72 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PIAS1(8554)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1O17 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects E3 SUMO-protein ligase PIAS1. It targets an epitope within the C-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 150 amino acids from the C-terminal region of human E3 SUMO-protein ligase PIAS1 protein.

Application

Anti-PIAS, clone 1O17 ZooMAb, Cat. No. ZRB1450, is a recombinant Rabbit monoclonal antibody that specifically targets PIAS and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PIAS in lysates from K562, L6, Sh-SY5Y, and NIH3T3 NE, and HeLa cell nuclear extracts.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PIAS in SH-SY5Y cells.

Affinity Binding Assay: A representative lot of this antibody bound recombinant E3 Sumo protein fragment with a KD of 4.7 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected PIAS in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

E3 SUMO-protein ligase PIAS1 (UniProt: O75925; also known as DEAD/H box-binding protein 1, E3 SUMO-protein transferase PIAS1, Gu-binding protein, GBP, Protein inhibitor of activated STAT protein 1, RNA helicase II-binding protein) is encoded by the PIAS1 (also known as DDXBP1) gene (Gene ID: 8554) in human. PIAS is an E3-type small ubiquitin-like modifier (SUMO) ligase that is expressed in numerous tissues with highest levels found in the testis. It stabilizes the interaction between UBE2I and the substrate and also acts as a SUMO-tethering factor. PIAS plays a crucial role as a transcriptional coregulation in various cellular pathways, including the STAT pathway, the p53 pathway and the steroid hormone signaling pathway. The effects of this transcriptional coregulation, transactivation or silencing, may vary depending upon the biological context. It is known to enhance the sumoylation of MTA1 and may also participate in its paralog-selective sumoylation. It also plays a role in adipogenesis by promoting the SUMOylation and degradation of CEBPB. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service