Skip to Content
MilliporeSigma
All Photos(11)

Documents

ZRB1222

Sigma-Aldrich

Anti-Beclin 1 Antibody, clone 1E22 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Coiled-coil myosin-like BCL2-interacting protein, Protein GT197

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E22, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 51.9 kDa
observed mol wt ~58 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BECN1(8678)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1E22 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Beclin-1. It targets an epitope within 24 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal region of human Beclin-1.

Application

Anti-Beclin 1, clone 1E22 ZooMAb, Cat. No. ZRB1222, is a highly specific rabbit recombinant monoclonal antibody that targets Beclin-1 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Beclin-1 in human breast cancer tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Beclin-1 in HeLa cells treated with Chloroquine (100 uM for 3 h).

Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary.

Target description

Beclin-1 (UniProt: Q14457; also known as Coiled-coil myosin-like BCL2-interacting protein, Protein GT197) is encoded by the BECN1 (also known as GT197) gene (Gene ID: 8678) in human. Beclin-1 is a peripheral membrane protein that plays a key role in autophagy. It is a component of the PI3K (PI3KC3/PI3K-III/class III phosphatidylinositol 3-kinase) complex the core of which is composed of the catalytic subunit PIK3C3, the regulatory subunit PIK3R4 and Beclin-1 associating with additional regulatory/auxiliary subunits to form alternative complex forms. It interacts with several cofactors, such as Atg14L, UVRAG, Rubicon, Ambra1, and survivin to regulate the activity of phosphatidylinositol 3-kinase. Its coiled-coil domain (aa 142-270) can form antiparallel homodimers and mediate dimerization with the coiled coil domains of ATG14 or UVRAG. Baclin-1 contains a BH3 motif (aa 108-127) that is shown to be bound to and inhibited by Bcl-2 or Bcl-xL. It also contains a C-terminal evolutionary conserved domain (ECD; aa 245-450) that has poly-Gln-binding domains and within this ECD it has a short amino acid sequence (aa 425-450) that is required for its membrane association. Beclin can undergo cleavage into Beclin-1-C (35 kDa; aa 134-450) and Beclin-1-C (37 kDa; aa 150-450). The Beclin-1-C 35 kDa localizes to mitochondria and can promote apoptosis by inducing the mitochondrial translocation of Bax and the release of proapoptotic factor. Beclin-1 is shown to undergo caspase-mediated cleavage that promotes crosstalk between apoptosis and autophagy. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service