Skip to Content
MilliporeSigma
All Photos(13)

Key Documents

ZRB1074

Sigma-Aldrich

Anti-DKK3 Antibody, clone 3H17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Dickkopf-3, Dickkopf-related protein 3, Dkk-3, hDkk-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H17, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 38.39 kDa
observed mol wt ~58 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DKK3(27122)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3H17 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Dickkopf-related protein 3 (DKK3).

Immunogen

His-tagged recombinant fragment corresponding to 300 amino acids (almost full length mature protein) from human Dickkopf-related protein 3 (DKK3).

Application

Affinity Binding Assay: A representative lot of this antibody bound recombinant DKK3 protein with a KD of 5.7 x 10-9 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected DKK3 in MCF-7 cells.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected DKK3 in one million MCF-7 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected DKK3 in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-DKK3, clone 3H17 ZooMAb, Cat. No. ZRB1074, is a recombinant Rabbit monoclonal antibody that targets DKK3 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Dickkopf-related protein 3 (UniProt: Q9UBP4; also known as Dickkopf-3, Dkk-3, hDkk-3) is encoded by the DKK3 (also known as REIC, UNQ258/PRO295) gene (Gene ID: 27122) in human. DKK is a family of secreted glycoproteins that display Wnt inhibitory activity. They act as high affinity ligands for the transmembrane proteins Kremen1 and 2, which are known to modulate Wnt signaling. They play a role in vertebrate development via locally inhibiting Wnt-regulated processes such as antero posterior axial patterning, limb development, somitogenesis and eye formation. DKK3 is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off in the mature form. High expression of DKK3 is reported in heart, brain, and spinal cord. DKK3 is shown to antagonize canonical Wnt signaling by inhibiting LRP5/6 interaction with Wnt and by forming a ternary complex with Kremen protein, which is known to promote internalization of LRP5/6. The C-terminal cysteine-rich domain of DKK3 mediates its interaction with LRP5 and LRP6. DKK3 is shown to induce a redistribution of β-catenin to the cell membrane where β-catenin promotes cell-cell adhesion by binding to the cytoplasmic domain of E-cadherin to form adheren junction complex. This binding also prevents nuclear translocation of β-catenin that reduces Wnt-mediated LEF/TCF transactivation. DKK3 is proposed to act as a tumor suppressor protein and its over-expression is shown to suppress tumor growth. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hoang, BH., et al. (2004). Cancer Res. 64(8); 2734-2339; Niehrs. C. (2006). Oncogene. 25(57); 7469-7481).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service