Skip to Content
MilliporeSigma
All Photos(14)

Documents

ZRB1051

Sigma-Aldrich

Anti-BCL-11A Antibody, clone 3B12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

B-cell CLL/lymphoma 11A, B-cell lymphoma/leukemia 11A, BCL-11A, COUP-TF-interacting protein 1, EVI-9, Ecotropic viral integration site 9 protein homolog, Zinc finger protein 856

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

3B12, recombinant monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 91.20 kDa

species reactivity

rat, human

species reactivity (predicted by homology)

bovine, sheep

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3B12 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects B-cell Lymphoma/leukemia 11 (BCL-11A). It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human B-cell Lymphoma/leukemia 11 (BCL-11A).

Application

Quality Control Testing
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected BCL-11A in human tonsil tissue sections.Tested applicationsImmunocytochemistry Analysis: A 1:100 dilution from a representative lot detected BCL-11A in HeLa and A431 cells.Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected BCL-11A in Ramos and Raji cell lysates.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-BCL-11A, clone 3B12 ZooMAb, Cat. No. ZRB1051, is a recombinant Rabbit monoclonal antibody that specifically targets BCL-11A and is tested for use in and Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

B-cell lymphoma/leukemia 11A (UniProt: Q9H165; also known as BCL-11A, B-cell CLL/lymphoma 11A, COUP-TF-interacting protein 1, Ecotropic viral integration site 9 protein homolog, EVI-9, Zinc finger protein 856) is encoded by the BCL11A (also known as CTIP1, EVI9, KIAA1809, ZNF856) gene (Gene ID: 53335) in human. BCL-11A is a transcription factor associated with the BAF SWI/SNF chromatin remodeling complex that also acts as a repressor of fetal hemoglobin levels and plays a role in hematopoiesis. It is expressed at high levels in brain, spleen thymus, bone marrow and testis. Its expression has also been reported in CD34+ myeloid precursor cells, B-cells, monocytes and megakaryocytes. Its expression is tightly regulated during B-cell development. BCL-11A contains six C2H2 type zinc-finger regions. It is found in the nucleus as well as in the cytoplasm. In the absence of interaction between isoforms 1 and 2, it is predominantly localized in the cytoplasm and localizes to the nuclear paraspeckles when both isoforms are present. Chromosomal aberrations involving BCL11A gene are reported in B-cell malignancies. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service