Skip to Content
MilliporeSigma
All Photos(7)

Documents

ZMS1188

Sigma-Aldrich

Anti-Perforin-1 Antibody, clone dG9 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cytolysin, Lymphocyte pore-forming protein, P1, PFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

dG9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 61.38 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone dG9 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Perforin-1.

Immunogen

Purified granules from the human cytotoxic large granular lymphocytes lymphoma line YT.

Application

Quality Control Testing

Evaluated by Western Blotting with His/SUMO-tagged recombinant Human Perforin-1.

Western Blotting Analysis: A 1:500 dilution of this antibody detected His/SUMO-tagged recombinant Human Perforin-1.

Tested Applications

Flow Cytometry Analysis: 1 µg from a representative lot detected Perforin-1 in one million Human peripheral blood mononuclear cells (PBMC).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Perforin-1 in SW480 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Perforin-1 (UniProt: P14222; also known as P1, Cytolysin, Lymphocyte pore-forming protein, PFP) is encoded by the PRF1 (also known as PFP) gene (Gene ID: 5551) in human. Perforin-1 is a multi-pass membrane glycoprotein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to produce the mature form that contains two transmembrane domains (aa 129-149 and 257-279). It is stored in cytolytic granules of cytolytic T-lymphocytes and is secreted into the cleft between T-lymphocyte and target cell. Perforin has a thin KEY®-shaped structure consisting of an N-terminal membrane attack complex perforin-like (MACPF)/ cholesterol dependent cytolysin (CDC) domain (aa 27-375), which is followed by an EGF domain (aa 376-408) that together with the eXTReme C-terminal sequence forms a central shelf-like structure.  Its C-terminal C2 domain (aa 397-519) mediates initial, Ca2+-dependent membrane binding. Perforin-1 serves as a pore-forming protein that plays a KEY® role in granzyme-mediated programmed cell death, and in defense against virus-infected or neoplastic cells. It also plays an important role in killing other cells that are recognized as non-self by the immune system. It can insert into the membrane of target cells in its calcium-bound form and can oligomerize and form large pores. Its membrane insertion and pore formation require a major conformation change. It promotes cytolysis and apoptosis of target cells by facilitating the uptake of cytotoxic granzymes. Mutations in PRF1 gene have been linked to familial Hemophagocytic lymphohistiocytosis (FHL2), a rare disorder that is characterized by immune dysregulation with hypercytokinemia, defective function of natural killer cell, and massive infiltration of several organs by activated lymphocytes and macrophages. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Praper, T., et al. (2011). J. Biol. Chem. 286(4); 2946-2955; Law, RHP., et al. (2010). Nature. 468(7322). 447-451; Ando, K., et al. (1997). J. Immunol. 158(11); 5283-5291).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
KEY is a registered trademark of Hanna Instruments
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany
eXTReme is a trademark of Excel Scientific, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service