Key Documents
Z763071
Eppendorf® Combitips advanced® assortment pack
1 of each of the 9 tip volumes and one adapter for the 25 and 50 mL tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 0030089936
maximum volume
50 mL
volume range
0.1-50 mL
General description
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
Legal Information
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)