Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z744281

Wilmad® Universal Jet Washer for EPR and NMR Sample Tubes

tube O.D. 2.5-5 mm, plain joint

Synonym(s):

EPR tube washer, NWR tube washer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121705
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-7200-1

tube O.D.

2.5-5 mm

Looking for similar products? Visit Product Comparison Guide

General description

Universal Jet NMR Tube Washer can be used for any length NMR sample tube by simply adjustmenting the included flexible PTFE tubing. The washer head is then affixed to a 24/40 joint filter flask (with vacuum sidearm) and the tubing is inserted into a washing solvent reservoir. An inverted sample tube is placed over the PTFE tubing, a vacuum is applied to the flask, sample tube is pressed against the rubber gasket to form an air-tight seal that starts the solvent flow. By lifting the PTFE tubing out of the solvent reservoir, the sample tube can be air-dried.

Application

NMR

Features and Benefits

For any length NMR sample tube

Legal Information

Corning is a registered trademark of Corning, Inc.
Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service