Documents
Z744248
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
offset for Bruker®, pack of 1 ea
Synonym(s):
EPR cryo dewar, EPR dewar
About This Item
UNSPSC Code:
41121804
NACRES:
NB.15
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-819-B-Q
General description
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir. The offset versions (WG-819) are designed for cavities that have limited space between the reservoir and the drive shaft for iris.
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries