Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z744195

eXTReme SealMate Adhesive Microplate-sealing Film Rolls

non-sterile

Synonym(s):

microplate sealing film, multiwell plate sealing film, sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410

sterility

non-sterile

feature

DNase free
RNase free

packaging

pkg of 160 ea (2 rolls, 80 sheets per roll)

manufacturer/tradename

Excel Scientific RSMXTR-2

parameter

-20-+120 °C temp. range

technique(s)

PCR: suitable

General description

Extra strong adhesive minimizes evaporation but removes without residue. These replacement rolls for the SealMate dispenser are cost effective. Replacement rolls can be easily installed by releasing the locking clasps on the dispenser

Application

PCR, General Storage

Features and Benefits

  • Extra strong adhesive minimizes evaporation yet removes easily without leaving residue on the plate.
  • Replacement rolls can be easily installed by releasing the locking clasps on the dispenser

Legal Information

SealMate is a trademark of Excel Scientific, Inc.
eXTReme is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service