Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z744117

Freezer box divider

to hold, 100 tubes, pk of 12

Synonym(s):

freezer container, freezer storage, storage container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804

feature

to hold 100 tubes

packaging

pack of 12
pk of 12

manufacturer/tradename

Diversified Biotech FBCD-4000

Looking for similar products? Visit Product Comparison Guide

General description

Freezer box divider, 100 hole 12/pk

Application

Cryogenics

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service