Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z744094

Planet-Safe® compostable weigh boat

W × L 3.6 in. × 3.6 in.

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign Into View Organizational & Contract Pricing

Select a Size

500 EACH
$166.00

$166.00


Please contact Customer Service for Availability


Select a Size

Change View
500 EACH
$166.00

About This Item

UNSPSC Code:
41121812

$166.00


Please contact Customer Service for Availability

packaging

pack of 500 ea

manufacturer/tradename

Diversified Biotech BOAT-3000

W × L

3.6 in. × 3.6 in.

General description

Made from a uniquely formulated bio-polymer, Weighing boats derive from 100% renewable resources. Weighing boats are anti-static and are “enviro-green” providing good visual contrast for powders. When finish, boats can be incinerated, composted (commercially), recycled (where accepted). Though it is least desirable, items disposed of in the trash will not release methane in a landfill. In all end-of-life scenarios, boats will breakdown into carbon neutral components.

Features and Benefits

Derived from 100% renewable resources. Good visual contrast for powders

Legal Information

Planet-Safe is a registered trademark of Diversified Biotech, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service