Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z744086

Solvent resistant pen

black

Synonym(s):

Pen

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$14.30

$14.30


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$14.30

About This Item

UNSPSC Code:
41103032
NACRES:
NA.21

$14.30


Please contact Customer Service for Availability

material

black

packaging

pack of 1 ea

manufacturer/tradename

Diversified Biotech 1000-BLACK

technique(s)

cryopreservation: suitable
electron microscopy: suitable
immunohistochemistry: suitable

General description

Resistant to most solvents including xylene, alcohol, and DMSO. May be used in the presence of acetone showing moderate resistance.  Ideal for histology cassettes, microscope slides and cryogenic vials. Markings will not run on frosted/painted labware. Markings are smear-proof, odorless and fade-resistant.

Application

Histology, Microscopy, Cryogenics

Features and Benefits

Resistent to most solvents including xylene, alcohol and DMSO.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is this pen suitable for labels and is it a thin tip pen?

    1 answer
    1. This product is a fine tip pen that is resistant to most solvents, such as xylene and DMSO. The marker is best suited for use with labware items such as histology cassettes, microscope slides and cryogenic vials. The marker is expected to be suitable for use with typical paper labels.

      Helpful?

  2. Would this be suitable for use in the same way as a PAP pen? Is it also water resistant and could be used to draw a hydrophobic barrier around a section

    1 answer
    1. This product is not intended to be used as a PAP pen. This pen is resistant to xylene, alcohols, and DMSO and shows moderate resistance to acetone. The item is generally used to label histology cassettes, microscope slides and cryogenic vials. Markings will not run on frosted/painted labware and are smear-proof, odorless and fade-resistant.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service