Z743662
Thermal Transfer Tough-Tags™
white, L × W 1.75 in. × 0.75 in., roll of 2000
Synonym(s):
solvent resistant labels, temperature resistant labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
packaging
roll of 2000
manufacturer/tradename
Diversified Biotech THER-1175
L × W
1.75 in. × 0.75 in.
color
white
Looking for similar products? Visit Product Comparison Guide
General description
- Temperature resistant from -40°c to 150°c
- Print popular barcode formats forfast and easy sample identification
- Print as little as one label at a time, eliminating label waste and saving time and money
- Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
- Resists degradation from scuffing, chemicals, moisture and wide temperature fluctuations
- Coated for use with a resin ribbon that provides the best performance under extreme conditions
- Works with most thermal transfer printers
Legal Information
Tough-Tags is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service