Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z741954

Cameo Syringe Filter

polyethersulfone membrane, pore size 0.45 μm, diam. 30 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

material

polyethersulfone membrane
polypropylene housing

sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

GVS Life Sciences 1233550

diam.

30 mm

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.

Applications:
Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis

Legal Information

Cameo is a trademark of GVS S.p.A.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service