Key Documents
Z741754
Manifold
2-Place
Select a Size
1 EA
$95.40
$95.40
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$95.40
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
$95.40
Please contact Customer Service for Availability
material
polysulfone
feature
autoclavable
Includes necessary tubing – 1/8” i.d. silicone
manufacturer/tradename
IBI ACCFL0005
General description
The manifold allows for the connection and simultaneous delivery of media to flow cells and for consolidation of effluent from cells to the waste bottle. Comes with 1/8” i.d. silicone tubing attached and is autoclavable and re-usable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)