Key Documents
Z740467
MICROGARD 1500 PLUS coveralls for protection from particules and light spray
size XXL
About This Item
UNSPSC Code:
46181503
NACRES:
NB.35
size
XXL
manufacturer/tradename
Arco 1763107
General description
MICROGARD 1500 PLUS white coverall is made from a highly breathable anti-static SMS fabric which utilises the latest developments in micro-fibre technology to ensure good filtration efficiency, offers protection from particles, perfect for working with asbestos, powders and general direct and dust with added light liquid spray protection.
Features:
  • 3 piece hood
  • 2-way front zipper with re-sealable storm flap
  • Anti-static treatment
  • Elasticated hood, wrists, waist and ankles
  • Stitched seams
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced Spray
EN1149-5 Anti-static
EN1073-2 Barrier to radioactive particulates - Class 1
Legal Information
Microgard is a trademark of Microgard Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)