Z740447
Eppendorf® Reference® 2 Variable Volume Pipettor
volume - 0.1–2.5 μL, 0.5–10 μL, 10–100 μL, 100–1,000 μL, pack of 4 ea
Synonym(s):
Eppendorf Variable Volume Pipette
About This Item
feature
electronic: no
single-channel
packaging
pack of 4 ea
manufacturer/tradename
Eppendorf® 2231302001
volume measuring range
0.1-2.5 μL
0.5-10 μL
10-100 μL
100-1000 μL
volume
1000 μL
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
General description
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service