Key Documents
Z740417
Specimen containers with snap cap
size 300 mL
Select a Size
100 EA
$185.00
$185.00
Please contact Customer Service for Availability
Select a Size
Change View
100 EA
$185.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
$185.00
Please contact Customer Service for Availability
material
polypropylene cap
polypropylene container
packaging
pack of 100 ea
manufacturer/tradename
Simport C580-3
size
300 mL
General description
These disposable containers made of polypropylene, are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting
Choose from one of the most recent versions:
Certificates of Analysis (COA)