The piece closest to the light source designed to collect and condense the light and then to focus it onto the specimen.
Z737712
Jenco™ compound microscopes
AC/DC input 220 V AC, brightfield, achromatic lens, binocular, mechanical stage; led or other illumination
Synonym(s):
compound microscopes, microscopes
About This Item
Recommended Products
description
3-plug
EU 2-prong plug
manufacturer/tradename
Jenco CP135-B4-2
General description
- Illumination can be a mirror, a rechargeable LED, an incandescent bulb or halogen bulb
- Eye pieces come in monocular heads, or teaching heads for assisting students in finding specimens and also binocular heads for easy viewing with both eyes
- Objectives are offered in multiple quality and quantity levels
- Video systems are available to project images on a screen
- Industry leading 5 year warranty
Choose from different frames for the right reason and teach science with the right equipment for your students.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
What is a Condenser on a Jenco microscope?
1 answer-
Helpful?
-
-
What does the working distance on a Jenco™ microscope refer to?
1 answer-
The working distance is measured from the end of the objective to the top of the specimen when the specimen is in focus.
Helpful?
-
-
What objectives can you use immersion oil with on a Jenco™ microscope?
1 answer-
Only those objectives designed for it with an oil specification clearly marked on it's barrel (100X OIL), otherwise the oil creeps up into the microscope causing problems.
Helpful?
-
-
Do Jenco™ microscopes have the option of increasing magnification through eyepieces?
1 answer-
Yes. All compound microscopes have a 15X option and the Stereo Zoom microscopes have a 15X and 20X option.
Helpful?
-
-
How do you use immersion oil on a Jenco™ microscopes?
1 answer-
Put a drop of oil on top of the slide's cover slip; move the objective down until it touches the oil and you have the specimen in focus.
Helpful?
-
-
Can you use a 35mm camera with your Jenco™ microscopes?
1 answer-
We do not suggest using a 35mm Camera as we have found that a digital camera provides a clearer picture.
Helpful?
-
-
What is a Stage or Mechanical stage on a Jenco™ microscope?
1 answer-
A flat plate for placing the slide/specimen with controls to easily move the sample both vertically and horizontally in small increments.
Helpful?
-
-
What is the difference between achromatic, semi-planachromatic, and planachromatic objectives on a Jenco™ microscope?
1 answer-
Each type of objective is color corrected to a higher degree than the previous one. In everyday terms, achromatic objectives afford approximately 80% of the field of view in focus from the center out to the edges. Semi-planachromatic afford approximately 90% and planachromatic afford approximately 100%. The better the color correction, the more cost.
Helpful?
-
-
What Jenco™ microscopes can be used wiith video or digital camera equipment?
1 answer-
We recommend only those models with trinocular heads because they are more stable and we have adapters to fit the trinocular tube, not the eyepiece, Jenco Model BC-311 (Sigma nos. Z735086 for 115V or Z737062 for 220V).
Helpful?
-
-
Do your Jenco™ microscopes come with rubber eyecups?
1 answer-
Yes.
Helpful?
-