Documents
Z722847
Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Rotor, 6 place for CYTO, swing out, for ROTOFIX 32A and UNIVERSAL 320
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
feature
to hold 6 x 1660, 1680 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1626
parameter
2,039 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Totafix 32A and Universal 320/320R
Related Categories
General description
Carriers not included.
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG