Z722499
Spectrafuge™ Mini-Centrifuge Replacement Rotors
0.2 mL strip tubes or individual 0.2 mL tubes
Synonym(s):
equipment replacement accessory
Select a Size
$69.90
Select a Size
About This Item
$69.90
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Labnet C1300-RTS
Labnet
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Spectrafuge mini-centrifuge
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service