Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z721360

Protein array buffers and reagents

Protein Array Buffer (2x) 10ml, 4/pk

Synonym(s):

protein microarray buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

sterility

non-sterile

packaging

pack of 4

manufacturer/tradename

Whatman 10485331

volume

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

Protein Arraying Buffer

  • Enhances long-term protein stability and molecular recognition activity
  • Enhances fluorescent signal from arrayed sample
  • Optimized for use on FAST Slides
Whatman® Protein Array Buffers and Reagents

Whatman protein array reagents have been optimized for use on FAST Slides. These reagents include the following (each sold separately):

  • Protein Arraying Buffer
  • Protein Array Blocking Buffer
  • Protein Array Wash Buffer

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service