Key Documents
Z708259
Foam plugs
size 46-50 mm
Synonym(s):
foam plug
About This Item
UNSPSC Code:
41100000
size
46-50 mm
General description
Reusable, nonabsorbent plugs provide a snug fit yet allow free airflow. Easily manipulate to fit flasks or tubes. Plugs spring back to original shape after repeated use. Autoclavable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)