Key Documents
Z707716
TPP® centrifuge tubes
capacity 50 mL, polypropylene
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Pricing and availability is not currently available.
material
cap
conical bottom tube
polypropylene
transparent tube
sterility
sterile
packaging
pack of 320 ea
manufacturer/tradename
TPP 91051
capacity
50 mL
General description
The TPP 50 mL and 15 mL Centrifuge Tubes are compatible with all standard centrifuges and their innovative design focuses on a tight seal, excellent transparency, clear graduations, and highly visible marking areas. The yellow screw cap closure on the TPP Centrifuge Tubes is tight and protected against overturning. The transparency of the tubes is excellent and the white marking areas on the top of the cap and side of the tube are highly visible. On the conical tubes, the distinct white graduations are continued to the bottom of the tube to accommodate small sample volumes. On the 500 mL tubes, the scale starts at 0.5 mL, and on the 15 mL tubes, the scale starts at 0.1mL.
Temperature Range for Polypropylene Tubes: -196°C to +121°C; Polystyrene Tubes: -40°C to +70°C; Maximum RCF for Polypropylene Tubes: 15500 × g*; Polystyrene Tubes: 1700 × g*.
* Depending on adapter and temperature
All Tubes are gamma Irradiated
Legal Information
TPP is a registered trademark of TPP Techno Plastic Products AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)