Key Documents
Z659835
Bandelin SONOREX Ultrasonic bath accessories
positioning lid for Z659800 and Z659045
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
material
stainless steel
feature
for 4 beakers
packaging
pack of 1 ea
manufacturer/tradename
Bandelin DE 514
availability
not available in USA and Canada
General description
When using chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath, use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
Features and Benefits
  • for holding insert cups
  • sound-absorbing
  • shortens the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)