Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z653950

Pyrex® glass centrifuge tube, ground socket

joint: ST/NS 24/20, tube capacity 30 mL, tube O.D. × L 28.5 mm × 115 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

borosilicate glass
clear tube

Joint

joint: ST/NS24/20

feature

RCF 3,000 × g

packaging

pack of 10

manufacturer/tradename

Bibby Pyrex BC24/C24R

availability

available only in EU

tube O.D. × L

28.5 mm × 115 mm

tube capacity

30 mL

General description

  • Borosilicate glass with precision ground Quickfit® socket to accept tapered stopper
  • Cylindrical shape
  • Each tube is marked with its weight to the nearest 0.1 g to enable tubes of similar weight to be selected from any batch
  • Can be centrifuged at RCF calues up to 3,000

Legal Information

PYREX is a registered trademark of Corning, Inc.
Quickfit is a registered trademark of SciLabware Limited

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Walsh et al.
Scientific data, 7(1), 380-380 (2020-11-13)
The Southern Ocean zooplankton community is diverse, yet most species are understudied, especially with respect to their overwinter feeding ecologies. Here we present body condition and trophic biomarker data (lipid content and stable isotopes of carbon and nitrogen) from 19
Toshio Kawanami et al.
International journal of pharmaceutics, 585, 119519-119519 (2020-06-15)
A local sustained-release drug delivery system, or depot, for intra-articular injection offers the opportunity to release a therapeutic agent directly to the joint with limited need for reinjection. A successful system would provide more consistent efficacy and minimize systemic side
Lu Yang et al.
Journal of the science of food and agriculture, 101(4), 1609-1617 (2020-09-03)
Astaxanthin ester (Asta-E) is used as functional nutraceuticals in many food products. Unfortunately, Asta-E utilization is currently limited owing to its chemical instability and low bioavailability. The purpose of this study is to investigate the promotion effect of oil matrixes
I Papachristou et al.
Biotechnology for biofuels, 14(1), 20-20 (2021-01-16)
Microalgae have attracted considerable interest due to their ability to produce a wide range of valuable compounds. Pulsed Electric Fields (PEF) has been demonstrated to effectively disrupt the microalgae cells and facilitate intracellular extraction. To increase the commercial viability of
Jessica K Beekman et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(3), 374-390 (2020-01-10)
Fatty acid esters of 3-monochloropropane-1,2-diol (3-MCPD) and glycidol are potentially carcinogenic and/or genotoxic processing contaminants that are formed during the process of edible oil refining. Because of their toxicological properties, the presence of these compounds in refined oils and foods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service