Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z653454

Hettich® centrifuge adapters

for ROTOFIX 32A and UNIVERSAL 320/320R, Adapter (rubber insert) for 100 mL tubes (44x100 mm)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

feature

Holds 100 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 0761

technique(s)

centrifugation: suitable

compatibility

for use with 1481, 1490, 1495 (bucket)

Looking for similar products? Visit Product Comparison Guide

General description

  • compatible with bucket/carriers 1381 (Z601543), 1390 and 1430

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

required but not provided


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service