Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z646695

BRAND® Transferpette® S starter kits

Midi

Synonym(s):

Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Single channel pipettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

single-channel

packaging

pack of 3 ea

manufacturer/tradename

BRAND 704791

volume

20 μL × 200 μL × 1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Contains a set of 3 Transferpette S, Digital adjustable volume, 3 filled Tip-Box N and 3 wall/rack mounts.
Includes Transferpette S models D-20, D-200 & D-1000

Features and Benefits

  • The Transferpette® S is a single channel high quality pipette.
  • Includes Transferpette S models D-20, D-200 & D-1000.
  • High degree of accuracy and precision.
  • Comfortable grip improved ergonomics and easy cleaning.
  • Color-coded pipette.
  • One-handed volume adjustment with brake to protect against accidental volume changes
  • Easy in-lab calibration adjustments without tools.
  • Visible Four-digit volume display.
  • Universal tip cone fits tips from most quality manufacturers.
  • Completely and repeatedly autoclavable at 121°C/250°F (20 minutes).

Other Notes

BR704791 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service