Z646571
BRAND® Transferpette® S pipette, adjustable single channel
volume 1000-10000 μL, D-10000
Synonym(s):
Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Single channel pipettes
About This Item
Recommended Products
feature
single-channel
subdivision 10 μL
packaging
pack of 1 ea
manufacturer/tradename
BRAND 704784
tip size
, 1000-10000 μL
volume
1000-10000 μL
Looking for similar products? Visit Product Comparison Guide
General description
The Transferpette S pipettes are the perfect manual pipettes for the most demanding applications in the lab.
They include all of the features requested by Life Science users: rugged construction, one-hand volume adjustment, complete autoclavability, highest precision, and Easy Calibration technology for convenient, long-term reliability.
- Familiar centrally placed pipetting key and separate tip ejector
- Ergonomic finger rest that adapts to your hand for a relaxed grip
- Easy, one-handed volume setting for right-or left-handed users
- Easy-to-read 4-digit volume display for highest precision
- Complete autoclavability at 121°C (2bar), acc. to DIN EN 285
- Color-coding for easy tip selection
- Corrosion-resistant piston and ejector for durability
- Easy Calibration technology provides calibration adjustments without tools, and a clear indication when the pipettes have been adjusted from factory settings
Includes: Transferpette S, Digital adjustable volume, DE-M marking, with performance certificate, wall/rack mount.
* Filter tips
Features and Benefits
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service