Key Documents
Z645443
Nichiryo E- Pack reloading system
volume range 0.1-10 μL, elongated, Reloading system, 10 x 96
About This Item
UNSPSC Code:
41121600
material
colorless
sterility
sterile
feature
barrier: no
packaging
pack of 960 ea
pack of 960
manufacturer/tradename
Nichiryo BMT-SSEP
availability
not available in (France)
maximum volume
10 μL
volume range
0.1-10 μL
General description
Universal fit pipette tips supplied in racks with a convenient hinged lid giving quick easy access.
  • Manufactured RNAse and DNAse free
  • Eco-friendly design provides racked tips with minimum plastic waste
  • Can be used stacked or as individual racks
  • Convenient hinged rack provides quick, easy access to tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)