Key Documents
Z640867
Rocking table Kick II
add-on plate
About This Item
UNSPSC Code:
12352200
feature
CE compliant
General description
Rocking table KICK II, thermostattable large size incubation platform suitable for various incubation and preparation techniques (e. g. ELISAs, immunodiffusion, immunoblotting, histochemistry, in-situ-hybridization), cell- and microbiological culture experiments and for the accelerated processing of staining/destaining procedures with electrophoretic gels and blotting membranes.
A rugged and robust heavy-duty lab instrument with loading capacity up to 5 kg and an effective area of 45 × 30 cm covered with an anti-slip support. Constructed for continuous operation. Electronic regulation of lifting frequency (10 - 100/min) with newly developed lifting technique (lifting angle 5°, lifting height 3 cm) performing a continuous light "kick" agitation to prevent gels and membranes from sticking to their containment support!
Choose from one of the most recent versions:
Certificates of Analysis (COA)