Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z629243

Vivacell 250 ultrafiltration concentrator membrane inserts

MWCO 30 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24
Pricing and availability is not currently available.

material

polycarbonate housing
polyethersulfone membrane

sterility

non-sterile

packaging

package of 5 ea

manufacturer/tradename

Sartorius VC2521

availability

not available in USA and Canada, ROW

parameter

4 bar max. pressure (60 psi)

W × H

116 mm × 235 mm

dead stop volume

600 μL

membrane area

40 cm2

tube size

250 mL , centrifuge bottle

working volume

250 mL

pore size

30 kDa MWCO

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service