Documents
Z617849
Greiner PS tube
capacity × diam. × L 14 mL × 18 mm × 95 mm
About This Item
UNSPSC Code:
41121706
NACRES:
NB.22
material
cap (Two-Position Vent Stopper)
clear tube
polystyrene
polystyrene tube
round bottom tube
round bottom
straight neck
sterility
sterile
feature
RNase and DNase free
neck thread push fit
packaging
pack of 750 ea
manufacturer/tradename
Greiner 191161
capacity × diam. × L
14 mL × 18 mm × 95 mm
General description
  • High transparency
  • Round bottom with two position vent stopper