Documents
Z601195
Hettich® MIKRO 120 centrifuge accessories
adapter for 0.5-0.8 mL tubes 8x45 mm
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
feature
Holds 0.5 - 0.8 mL tubes
packaging
pack of 24 ea
manufacturer/tradename
Hettich 2023
technique(s)
centrifugation: suitable
compatibility
for use with 1213-A, 1226-A, 1252-A, 1258-A, 2428, 2434, 2437 (rotors)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG