Key Documents
Z379255
Sigma-Aldrich
Freezebee ice bucket insert
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
General description
Fits standard sized ice bucket to keep samples cold and prevent their being lost in the bed of ice. Ingenious pattern permits use with 0.5 mL (48 each) or 1.5 mL (47 each) microcentrifuge tubes, 15 mL (8 each) or 50 mL (6 each) conical tubes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)