Documents
Z376779
Sigma-Aldrich
Lazy-L Spreader
10 per pouch
Synonym(s):
L shape spreader, bacterial spreader, disposable cell spreader
About This Item
UNSPSC Code:
41161502
NACRES:
NB.22
material
polystyrene
sterility
sterile
packaging
case of 500 10 per pack
manufacturer/tradename
Excel Scientific SPR-L-S10
Lazy-L Spreader
General description
Lazy-L-Spreaders disposable microbiological spreaders provide an economical alternative to bending glass rods or pipets for spreading samples on agar surfaces and filters in Petri dishes. They are supplied sterile -- no need to flame sterilize or autoclave.
  • unlike glass spreaders, durable polystyrene Lazy-L spreaders are non-breakable and pose no safety hazard
  • L-shaped for convenient "lazy" spreading; just turn the Petri dish for smooth, even sample distribution
  • smooth spreading surface and a slight upward turn reduce the chance that the spreader will dig into agar
  • presterilization eliminates flaming or autoclaving and reduces risk of contamination
  • packaged in presterilized pouches of one or ten each
  • no more time spent bending glass rods or waiting for glass or metal spreaders to cool before use!
Legal Information
Lazy-L Spreader is a trademark of Excel Scientific, Inc.
Visit the Document Library
Articles
General protocols for growth of competent cells and their transformation (uptake of DNA).