Key Documents
Z374407
Sigma-Aldrich
Scienceware® microscope slide holder
accepts up to 10 single or double slides standing on end
Select a Size
1 EA
$33.70
$33.70
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$33.70
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
$33.70
Please contact Customer Service for Availability
feature
Holds 10 x slides 76x26 mm
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F44542-0000
D × W × H
10 in. (25 cm) × 2 in. (5 cm) × 1 1/4 in. (3 cm)
General description
Slide holder has a sloped groove that accepts up to 10 single or double slides standing on end. Made of durable, extruded white plastic,
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)