Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z363391

Sigma-Aldrich

Scienceware® replica plater for nutrient plate

pkg of 1 ea

Synonym(s):

culture replicator

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$251.00

$251.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$251.00

About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

$251.00


Please contact Customer Service for Availability

sterility

non-sterile

packaging

pkg of 1 ea

manufacturer/tradename

Bel-Art F37848-0000

color

none

General description

For transferring microbial colonies from one nutrient plate to others. The petri dish containing the colonies is inverted and lowered into contact with the velveteen, then removed. When fresh petri dishes, perhaps supplemented with antibiotics or lacking certain metabolites, are brought into contact with the velveteen, colonies are seeded onto the surface in the same pattern as the source plate.

Disinfect between uses by rinsing briefly with alcohol or 1:200 chlorine bleach. Designed for use with petri dishes from 90 mm to 100 mm. Dimensions: ring is 102 mm diameter; PVC cylinder is 69 mm high. Includes one bag of 12 velveteen squares.

Application

Sterile cotton velveteen square (150 × 150 mm) is draped over a solid PVC cylinder and locked in place with an aluminum ring.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Replica plating and indirect selection of bacterial mutants.
J LEDERBERG et al.
Journal of bacteriology, 63(3), 399-406 (1952-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service