Key Documents
Z359769
Peptide Antigens: A Practical Approach
About This Item
ISBN-10:
0-19-963451-3
ISBN-13:
978-0-19-963451-4
UNSPSC Code:
55101509
Pricing and availability is not currently available.
publication info
G.B. Wisdom, ed., IRL Press at Oxford University Press, Oxford, England, 1994, 252 pp., soft cover
manufacturer/tradename
Oxford University Press
Features and Benefits
This volume provides practical guidance to the major techniques used in the exploitation of peptide antigens and anti-peptide antibodies. The chapters give detailed protocols for the prediction of epitopes, peptide synthesis, the preparation of peptide immunogens, immunoaffinity chromatography, immunoassays, and the mapping of epitopes using both synthetic peptides and phage display systems.
Choose from one of the most recent versions:
Certificates of Analysis (COA)