Z328669
BRAND® magnetic stirring bar, PTFE, cylindrical
L 10 mm, bar diam. 6 mm
Select a Size
$66.90
Available to ship onApril 17, 2025Details
Select a Size
About This Item
$66.90
Available to ship onApril 17, 2025Details
Recommended Products
material
PTFE-coated
cylindrical
manufacturer/tradename
BRAND 137113
L
10 mm
bar diam.
6 mm
Related Categories
General description
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)