Key Documents
Z324892
Sigma-Aldrich
BRAND® SILBERBRAND ETERNA volumetric pipettes, calibrated to deliver
capacity 100 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
AR-Glas®
Agency
according to DIN 12 690
feature
class B (callibrated to deliver)
class B
one-mark
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29519
volume measuring range
100 mL, accuracy: 0.12 mL
capacity
100 mL
General description
The BRAND® SILBERBRAND ETERNA volumetric pipettes are expertly crafted fromAR-Glas® and conform to DIN EN ISO 648 standards, ensuring high precision and reliability. Calibrated to deliver (TD, Ex), these pipettes feature a waiting time of 5 seconds for optimal accuracy in liquid transfer. Each BLAUBRAND® bulb pipette is supplied with a batch number and includes one batch certificate per packing unit from the manufacturer, guaranteeing traceability and quality. This combination of quality materials and thorough documentation makes these pipettes an excellent choice for demanding laboratory environments.
Features and Benefits
  • BRAND® SILBERBRAND ETERNA, 1-mark, volumetric pipette is used for the accurate measurement of instruments for precision analyses.
  • Calibrated to deliver (TD, Ex).
Other Notes
BR29519 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.