Key Documents
Z324876
Sigma-Aldrich
BRAND® SILBERBRAND ETERNA volumetric pipettes, calibrated to deliver
capacity 25 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
AR-Glas®
Agency
according to DIN 12 690
feature
class B (callibrated to deliver)
class B
one-mark
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29515
volume measuring range
25 mL, accuracy: 0.04 mL
capacity
25 mL
General description
The BRAND® SILBERBRAND ETERNA volumetric pipettes are expertly crafted fromAR-Glas® and conform to DIN EN ISO 648 standards, ensuring high precision and reliability. Calibrated to deliver (TD, Ex), these pipettes feature a waiting time of 5 seconds for optimal accuracy in liquid transfer. Each BLAUBRAND® bulb pipette is supplied with a batch number and includes one batch certificate per packing unit from the manufacturer, guaranteeing traceability and quality. This combination of quality materials and thorough documentation makes these pipettes an excellent choice for demanding laboratory environments.
Other Notes
BR29515 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.