Key Documents
Z315907
Eppendorf® Reference® pipette, fixed volume
volume 500 μL
About This Item
UNSPSC Code:
41121500
material
blue control button
description
fully autoclavable
feature
electronic: no
single-channel
manufacturer/tradename
Eppendorf® 4900 000.516
volume measuring range
500 μL
General description
Ejector sleeve removable, for easy access to narrow vessels.
Single button control of filling, blow-out and tip ejection.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)