Skip to Content
MilliporeSigma
All Photos(1)

Documents

WHAWB120112

QIAcard FTA Wash Buffer

Nucleic acid purification buffer, bottle size 25 mL

Synonym(s):

QIAcard FTA Wash Buffer (25 ml), Whatman FTA, Whatman FTA Purification Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.52

feature

Nucleic acid purification buffer

packaging

pack of 1 ea

manufacturer/tradename

Qiagen WB120112

bottle size

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

QIAcard FTA Wash Buffer is used for purifying nucleic acids captured and stored on FTA cards. This versatile reagent is recommended for use with QIAcard FTA and Indicating FTA Card formats, and EasiCollect formats.

Features and Benefits

  • This reagent can be used to wash punches in the "punch-in" procedure for standard STR amplification.
  • QIAcard FTA Wash Buffer allows for excellent results by removing PCR inhibitors and contaminants that can lower PCR yields.

Other Notes

Field of Use : For internal research use only

Legal Information

FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service