Skip to Content
MilliporeSigma
All Photos(2)

Documents

WH0245972M1

Sigma-Aldrich

Monoclonal Anti-ATP6V0D2 antibody produced in mouse

clone 7A4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ATP6D2, Anti-ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2, Anti-FLJ38708, Anti-VMA6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7A4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

ATPase H+ transporting V0 subunit d2 (ATP6V0D2) is expressed on the cell surface of kidneys and osteoclasts. The gene encoding it is localized on human chromosome 8q21.

Immunogen

ATP6V0D2 (NP_689778, 238 a.a. ~ 306 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ETLYPTFGKLYPEGLRLLAQAEDFDQMKNVADHYGVYKPLFEAVGGSGGKTLEDVFYEREVQMNVLAFN

Biochem/physiol Actions

ATPase H+ transporting V0 subunit d2 (ATP6V0D2) takes part in H+ translocation. It also has an important role in the maturation of osteoclasts and development of bones.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taesoo Kim et al.
Biochemical and biophysical research communications, 390(3), 585-590 (2009-10-13)
Bone homeostasis is tightly regulated by matrix-producing osteoblasts and bone-resorbing osteoclasts. During osteoclast development, mononuclear preosteoclasts derived from myeloid cells fuse together to form multinucleated, giant cells. Previously, we reported that the d2 isoform of the vacuolar (H(+)) ATPase V0
Annabel N Smith et al.
Journal of bioenergetics and biomembranes, 40(4), 371-380 (2008-08-30)
The multi-subunit vacuolar-type H(+)-ATPase consists of a V(1) domain (A-H subunits) catalyzing ATP hydrolysis and a V(0) domain (a, c, c', c", d, e) responsible for H(+) translocation. The mammalian V(0) d subunit is one of the least-well characterized, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service