Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0170850M1

Sigma-Aldrich

Monoclonal Anti-KCNG3 antibody produced in mouse

clone 5H2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-KV10.1, Anti-KV6.3, Anti-potassium voltage-gated channel, subfamily G, member 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$539.00

$539.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$539.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$539.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNG3(170850)

Related Categories

General description

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit functioning as a modulatory molecule. Alternative splicing results in two transcript variants encoding distinct isoforms. (provided by RefSeq)

Immunogen

KCNG3 (NP_579875, 23 a.a. ~ 121 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SRELLKDFPLRRVSRLHGCRSERDVLEVCDDYDRERNEYFFDRHSEAFGFILLYVRGHGKLRFAPRMCELSFYNEMIYWGLEGAHLEYCCQRRLDDRMS

Biochem/physiol Actions

Potassium voltage-gated channel, subfamily G, member 3 (KCNG3) has been studied to have a regulatory function. It interacts with the potassium channel Kv2.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yorikata Sano et al.
FEBS letters, 512(1-3), 230-234 (2002-02-20)
We report identification and characterization of Kv6.3, a novel member of the voltage-gated K(+) channel. Reverse transcriptase-polymerase chain reaction analysis indicated that Kv6.3 was highly expressed in the brain. Electrophysiological studies indicated that homomultimeric Kv6.3 did not yield a functional
L A Pardo et al.
Current medicinal chemistry, 19(5), 675-682 (2011-12-30)
K(V)10.1 has recently become generally accepted as a promising cancer target, as it is ectopically expressed in the majority of solid tumors. Due to its cell-surface accessibility, K(V)10.1 has a strong potential for tumor treatment and diagnosis. Given that its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service