Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0055624M7

Sigma-Aldrich

Monoclonal Anti-POMGNT1 antibody produced in mouse

clone 6C12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp761B182, Anti-FLJ20277, Anti-GnTI.2, Anti-MEB, Anti-MGAT1.2, Anti-protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POMGNT1(55624)

General description

The protein encoded by this gene is a type II transmembrane protein that resides in the golgi. It participates in O-mannosyl glycosylation, and is specific for alpha linked terminal mannose. Mutations in this gene are associated with muscle-eye-brain (MEB) disease. Alternatively spliced transcript variants have been found for this gene

Immunogen

POMGNT1 (NP_060209, 221 a.a. ~ 318 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FGEKHSKSPALSSWGDPVLLKTDVPLSSAEEAECHWADTELNRRRRRFCSKVEGYGSVCSCKDPTPIEFSPDPLPDNKVLNVPVAVIAGNRPNYLYRM

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Luz Uribe et al.
Molecular vision, 22, 658-673 (2016-07-05)
The POMGNT1 gene, encoding protein O-linked-mannose β-1,2-N-acetylglucosaminyltransferase 1, is associated with muscle-eye-brain disease (MEB) and other dystroglycanopathies. This gene's lack of function or expression causes hypoglycosylation of α-dystroglycan (α-DG) in the muscle and the central nervous system, including the brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service