Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0023759M2

Sigma-Aldrich

Monoclonal Anti-PPIL2 antibody produced in mouse

clone 2E11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CYC4, Anti-CYP60, Anti-MGC33174, Anti-MGC787, Anti-hCyP60, Anti-peptidylprolyl isomerase (cyclophilin)-like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPIL2(23759)

General description

This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. This protein interacts with the proteinase inhibitor eglin c and is localized in the nucleus. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

PPIL2 (AAH00022, 1 a.a. ~ 527 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGKRQHQKDKMYITCAEYTHFYGGKKPDLPQTNFRRLPFDHCSLSLQPFVYPVCTPDGIVFDLLNIVPWLKKYGTNPSNGEKLDGRSLIKLNFSKNSEGKYHCPVLFTVFTNNTHIVAVRTTGNVYAYEAVEQLNIKAKNFRDLLTDEPFSRQDIITLQDPTNLDKFNVSNFYHVKNNMKIIDPDEEKAKQDPSYYLKNTNAETRETLQELYKEFKGDEILAATMKAPEKKKVDKLNAAHYSTGKVSASFTSTAMVPETTHEAAAIDEDVLRYQFVKKKGYVRLHTNKGDLNLELHCDLTPKTCENFIRLCKKHYYDGTIFHRSIRNFVIQGGDPTGTGTGGESYWGKPFKDEFRPNLSHTGRGILSMANSGPNSNRSQFFITFRSCAYLDKKHTIFGRVVGGFDVLTAMENVESDPKTDRPKEEIRIDATTVFVDPYEEADAQIAQERKTQLKVAPETKVKSSQPQAGSQGPQTFRQGVGKYINPAATEQQRKSPQPVPLSPCPRRSPVGVLGTSAPGSSRLPDDH

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service